Learn More
BTX™ AgilePulse™ MAX Safety Stand for Flatpack
Product Code. 15477380
Change view
Click to view available options
Connection Type:
BNC Cable
Unit Size:
Each
Product Code. Connection Type Unit Size
15477380 BNC Cable Each
1 options
Product Code. 15477380 Supplier BTX™ Supplier No. 470209
  • ONLINE EXCLUSIVE PRICE
    £2838.68
    £2631.22 / Pack of 1
    Save £207.46
    7% Off
Estimated Shipment: 22-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
AgilePulse™ MAX Safety Stand for Flatpack for use with BTX™ AgilePulse™ MAX.
  • Cuvette module plugs directly into generator
  • Holds one cuvette
  • Safety swing shield secures cuvette in holder
Specifications
For Use With (Application) Electroporation
For Use With (Equipment) AgilePulse™ MAX, ECM™ 2001+
Product Type Safety Stands
Voltage 500 to 1000 V
Description AgilePulse MAX Safety Stand for Flatpack
Display Touchscreen
Includes Safety Stand Only
Sterility Non-sterile
Connection Type BNC Cable
Interface Digital User
Pulse Form AC Sine Wave, DC Square Wave