Learn More
Tenak™ Safety Spring Lock for Cryoboxes
Product Code. p-9281127 Shop All Tenak Products
Click to view available options
Size:
10/Pk.
100/Pk.
Unit Size:
Pack of 10
Pack of 100
Product Code. 15271004
Brand: Tenak™ TE76010
Only null left
Add to basket
Add to basket
Safety Spring Lock is used to prevent the cryoboxes from sliding out when pulling the rack in and out of the freezer.
  • Use this spring instead of the locking rod
  • Simply place the Spring lock on the lid of the cryobox and push the cryobox in the rack
  • The Spring will come out together with the cryobox when pulling it out
  • Can be used when there is a gab between the cryobox and the shelf from 1 to 6 mm
  • Can be used with TENAK Eco AluRack or Standard racks for upright freezer and Classic or EcoAlu racks for chest freezers, or with any other rack brand with a similar design
  • Can withstand temperatures below -200°C
Specifications
For Use With (Equipment) Cryoboxes
Size 10/Pk.
Type Spring Lock