missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Safety Pipetting Ball

Product Code. 10777964 Shop All Bürkle Products
Change view
Click to view available options
Color:
Blue
Red
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10777964 Blue Each
10496263 Red Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10777964 Supplier Buerkle™ Supplier No. 75540005
  • ONLINE EXCLUSIVE PRICE
    £17.60
    £17.23 / Pack of 1
    Save £0.37
    2% Off
View all promotions
In Stock
Add to basket
Add to basket
Bulk Quote Request

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Peleus ball, simple pipetting aid. Evacuation takes place via an automatic valve. By pressing the valves, the liquid is drawn up, dispensed or purged. The removable automatic valve also enables effective cleaning of the inside

  • For all pipettes up to 100 mL
  • Available in various colours
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Color Blue
Disposable Reusable
For Use With Pipette
Grip Type Round
Material Natural Rubber
Number of Channels 2
Sterility Non-sterile
Type Ball
For Use With (Equipment) up to 100 mL Pipettes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.