missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Safety Pipetting Ball

Product Code. 10787964 Shop All Bürkle Products
Change view
Click to view available options
Color:
Green
Yellow
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10787964 Green Each
10580303 Yellow Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10787964 Supplier Buerkle™ Supplier No. 75540006
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Buerkle™ Safety Pipetting Ball allows evacuation to take place via an automatic valve.

  • For all pipettes up to 100 ml
  • Available in various colours
TRUSTED_SUSTAINABILITY

Specifications

Color Green
Type Ball
For Use With (Application) Enables effective cleaning of the inside
For Use With (Equipment) up to 100 mL Pipettes
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.