missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Edmund Buehler™ Safety package: Monitoring of Shaker Speed
Click to view available options
For Use With (Equipment):
KS 15 A Control, KS 15 B Control
SM 30 A Control, SM 30 B Control, SM 30 C Control, SM 30 AT Control
TiMix 5 Control
VKS 75 A Control, VKS 75 B Control
Unit Size:
Each
Description
- In case of deviations, e.g. due to a defective drive belt, a warning message is displayed and an acoustic signal is audible
- Monitoring of shaker speed
- In case of deviations, a warning message is displayed and an acoustic signal is audible
Specifications
Specifications
| Product Type | Safety Package |
| For Use With (Equipment) | KS 15 A Control, KS 15 B Control |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction