missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Safety-Labeled Wide-Mouth Wash Bottles
Shop All SP Scienceware Products
Click to view available options
Chemical Label:
Acetone
Distilled Water
Isopropanol
Sodium Hypochlorite (Bleach)
Description
- Wide-mouth (53 mm / 2 in.) design allows easy refilling and handling.
- Pre-printed labeling includes the chemical name and formula, NFPA Section 704 four-color diamond, health hazards, CAS number, and recommended protective equipment.
- Made of translucent LDPE; Sodium Hypochlorite bottle is white LDPE for light protection (230 - 450 nm), and Toluene bottle is red LDPE.
- Color-coded polypropylene closures ensure quick and easy chemical identification.
- Complies with OSHA Hazard Communication Standard 29 CFR 1910.1200.
- Available in 500 ml (4 per bag) and 1000 ml (4 per bag); assorted set includes six bottles - one each of Acetone, Bleach, Distilled Water, Ethanol, Isopropanol, and LYOB.
Note: Use LYOB only with solvents compatible with LDPE.
Specifications
Specifications
| Capacity (Metric) | 500 ml |
| Capacity (English) | 16 oz. |
| Color | White |
| Material | LDPE |
| No. per Pack | 4/Pk. |
| Sterility | Non-sterile |
| Closure Color | Yellow |
| Closure Included | Yes |
| Disposable | Reusable |
| Graduated | No |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction