missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Safety-Labeled Wide-Mouth Wash Bottles
Description
- Wide-mouth (53 mm / 2 in.) design allows easy refilling and handling.
- Pre-printed labeling includes the chemical name and formula, NFPA Section 704 four-color diamond, health hazards, CAS number, and recommended protective equipment.
- Made of translucent LDPE; Sodium Hypochlorite bottle is white LDPE for light protection (230 - 450 nm), and Toluene bottle is red LDPE.
- Color-coded polypropylene closures ensure quick and easy chemical identification.
- Complies with OSHA Hazard Communication Standard 29 CFR 1910.1200.
- Available in 500 ml (4 per bag) and 1000 ml (4 per bag); assorted set includes six bottles - one each of Acetone, Bleach, Distilled Water, Ethanol, Isopropanol, and LYOB.
Note: Use LYOB only with solvents compatible with LDPE.
Specifications
Specifications
| Capacity (Metric) | 500 ml |
| Capacity (English) | 16 oz. |
| Color | Clear |
| Material | Polyethylene |
| No. per Pack | 4/Pk. |
| Sterility | Non-sterile |
| Closure Color | Red |
| Closure Included | Yes |
| Disposable | Reusable |
| Graduated | No |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?