missing translation for 'onlineSavingsMsg'
Learn More

Uvex Safety Gloves U-Fit 60168

Product Code. p-200104594 Shop All Uvex Products
Click to view available options
Size:
Medium
Large
X-Large
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 30216655

Brand: Uvex 6016808

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The Uvex U-Fit series of high-quality disposable safety gloves are made of NBR and ensure a high level of safety and functionality.

  • High-quality disposable safety gloves
  • Made of NBR
  • High level of safety and functionality
  • Reliable protection
  • Comfortable
  • Precise working in various application
TRUSTED_SUSTAINABILITY

Specifications

Applications Healthcare, Laboratory, Industrial
Color Blue
Finger Thickness (English) 3 mil.
Finger Thickness (Metric) 0.08 mm
Latex Free Yes
Material NBR
Palm Thickness (English) 3 mil.
Palm Thickness (Metric) 0.08 mm
Product Type Chemical Protection Gloves
Size Medium
Thickness (English) 3 mil.
Thickness (Metric) 0.08 mm
EN ISO 374-1 Type A
EN ISO 374 - 5 Bacteria, Fungi only
Lining Unlined
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.