Learn More
uvex Safety Gloves HexArmor S6
Product Code. p-200115206 Shop All Uvex Products
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
1 pair
Product Code. 30533571
Brand: uvex 6066806
HexArmor™ safety gloves are available in a variety of sizes and provide protection against cuts and pinpricks.
HexArmor™ brand products stand for exceptional protection.
  • The exclusive SuperFabric™ brand material offers pinprick protection and extremely high cut protection.
  • For exceptional impact protection, the patented IR-X™ Impact Exoskeleton™ is an attractive solution.
TRUSTED_SUSTAINABILITY
Specifications
Construction Seamless Knit
Coating Material Polyurethane
Color Blue, Light Blue
Cut Resistance Rating ANSI A6
Hand Ambidextrous
Latex Free Yes
Material SuperFabric™
Product Type Cut-resistant Gloves
Size 6
Gender Unisex
Glove Cuff Style Knitwrist
Grip Design Smooth
Lining Seamless Lining
Packaging Type Pair
Puncture Rating ASTM F2878 LEVEL 4
Thumb Style Straight