Learn More
uvex Safety Gloves HexArmor S6
Product Code. 30533571 Shop All Uvex Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
1 pair
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
30533571 6 1 pair
30533599 7 1 pair
30533593 8 1 pair
30533600 9 1 pair
30533570 10 1 pair
30533589 11 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 30533571 Supplier uvex Supplier No. 6066806
  • ONLINE EXCLUSIVE PRICE
    £9.30
    £8.62 / 1 pairs
    Save £0.68
    7% Off
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
HexArmor™ safety gloves are available in a variety of sizes and provide protection against cuts and pinpricks.
HexArmor™ brand products stand for exceptional protection.
  • The exclusive SuperFabric™ brand material offers pinprick protection and extremely high cut protection.
  • For exceptional impact protection, the patented IR-X™ Impact Exoskeleton™ is an attractive solution.
TRUSTED_SUSTAINABILITY
Specifications
Construction Seamless Knit
Coating Material Polyurethane
Color Blue, Light Blue
Cut Resistance Rating ANSI A6
Hand Ambidextrous
Latex Free Yes
Material SuperFabric™
Product Type Cut-resistant Gloves
Size 6
Gender Unisex
Glove Cuff Style Knitwrist
Grip Design Smooth
Lining Seamless Lining
Packaging Type Pair
Puncture Rating ASTM F2878 LEVEL 4
Thumb Style Straight