missing translation for 'onlineSavingsMsg'
Learn More

TerraCycle™ Safety Equipment and Protective Gear Zero Waste Box™

Product Code. 17264934
Change view
Click to view available options
Capacity (Metric):
28 L
78 L
144 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric)
17264934 144 L
17284934 28 L
17274934 78 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17264934 Supplier TerraCycle™ Supplier No. CS58LUK
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recycle Safety Equipment and Protective Gear using TerraCycle's Zero Waste Box system. Zero Waste Box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.

WHAT YOU CAN RECYCLE

Use this Zero Waste Box to recycle:

  • Earplugs
  • Beard nets
  • Hairnets
  • Disposable gloves
  • Safety glasses
  • Disposable garments
  • Disposable shoe covers
  • Disposable masks*

*Accepts all types of disposable face masks made from plastic, such as dust masks, FFP2 (N95), FFP3, and surgical masks (as long as they don't come from a medical environment).

Do not send in:

  • Medical waste. This includes material contaminated with blood or bodily fluids and waste that originate from health care facilities, hospitals, home care, physicians' offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.
  • Fabric, homemade or paper face masks
  • Wax earplugs
  • Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) or material contaminated with blood which present a danger to the environment, or to people
  • Batteries, pressurised canisters, broken glass
  • Lateral flow test kits

Includes:

  • One Zero Waste Box
  • One prepaid return label, affixed to your box
  • Processing and recycling of the materials
  • Customer support from our team of experts
  • Important Shipping Restrictions

This service is only available in mainland Britain.

We don't operate in Northern Ireland, British Overseas Territories, Crown Dependencies (Guernsey Island, Jersey Island, the Isle of Man), the Channel Islands, the Hebrides/Shetland/Orkney Islands and the Isles of Scilly.

Zero Waste Box Shipping Notes:

For the Zero Waste Box system to work effectively and safely, please follow these simple shipping guidelines on how and what you can ship back to TerraCycle:

  1. Only send in items this system accepts. Sending in items that fall outside the selected category causes contamination and the need for added separation, which increases costs. This includes (but not limited to) liquid and food waste, cardboard, paper, compostable material such as PLA and PHA, mixed fibre fabric, borosilicate glass and electronic waste.
  2. Do not send in household items that may be considered hazardous waste. These include items such as lithium-ion batteries, pressurized canisters, pesticides, oil-based paint or medical sharps, organics, broken glass, biomedical waste, soiled nappies, or expired medication. Do not send in anything sharp, flammable, reactive, corrosive, ignitable, radioactive, toxic, infectious, or pathogenic.
  3. Do not ship dripping packages. Please make sure all excess product has been removed before putting it in your Zero Waste Box. If you choose to rinse your items, please note that the system must be completely dry prior to shipping.

Specifications

Capacity (Metric) 144 L
Certifications/Compliance ISO 9001:2015 Certified
Color White, Green
Disposable Single-use
Height (Metric) 97 cm
Latex Free Yes
Length (Metric) 38 cm
Sterility Non-sterile
Width (Metric) 38 cm
Autoclavable Not Autoclavable
Liner Material Plastic
For Use With (Application) Recycling
Material Cardboard
Product Type Recycling Box
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.