missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAT™ Safety Canisters, HDPE-EC
Shop All SCAT Europe Products
Click to view available options
Capacity (Metric):
5 L
10 L
20 L
30 L
60 L
Closure Size:
S50
S60/61
S70/71
S90
Includes:
Canister
Canister with Blue Stripes on Grip, without UN-Y Approval
Canister with Floater
Canister with Green Stripes on Grip, without UN-Y Approval
Canister with Red Stripes on Grip, without UN-Y Approval
Canister with UN-Y Approval
Canister with Viewing Strip, UN-Y Approval
Canister with Yellow Stripes on Grip, without UN-Y Approval
Unit Size:
Each
Description
- Made of very robust and electrically conductive HDPE, which is very resistant to solvents commonly used in laboratories
- Filling quantities from 10 L for use with HPLC systems up to massive 60 L drums for the collection of large quantities of containers (e.g. professional SymLine™ disposal systems)
- Thread types S50, S60/61, S70/71 and S90 are commonly used in the laboratory and are all compatible
- Safety Waste Caps range for the safe disposal of volatile, hazardous substances
- Special safety features such as integrated visibility strips or floats ensure permanent fill level monitoring
- Practical designs as space-saving canisters allow safe storage of liquid waste even in small areas or in hazardous goods cabinets
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Capacity (English) | 5.3 gal. |
| Capacity (Metric) | 20 L |
| Certifications/Compliance | UN-Y approval |
| Closure Size | S60/61 |
| Color | Black |
| Description | Canister, 20 Liter, S60/61, PE-HD electrostatic conductive, UN-Y approval, dimensions (W x H x D): 290 x 399 x 245 mm |
| Dimensions (L x W x H) | 290 x 399 x 245 mm |
| For Use With (Application) | Collection of Flammable Liquid Waste |
| Includes | Canister with UN-Y Approval |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction