missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Made of very robust and electrically conductive HDPE, which is very resistant to solvents commonly used in laboratories
- Filling quantities from 10 L for use with HPLC systems up to massive 60 L drums for the collection of large quantities of containers (e.g. professional SymLine™ disposal systems)
- Thread types S50, S60/61, S70/71 and S90 are commonly used in the laboratory and are all compatible
- Safety Waste Caps range for the safe disposal of volatile, hazardous substances
- Special safety features such as integrated visibility strips or floats ensure permanent fill level monitoring
- Practical designs as space-saving canisters allow safe storage of liquid waste even in small areas or in hazardous goods cabinets
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Capacity (English) | 1.63 gal. |
| Capacity (Metric) | 5 L |
| Certifications/Compliance | No UN-Y approval |
| Closure Size | S50 |
| Color | Black |
| Description | Space saving canister, 5 L, S50, PE-HD electrostatic conductive, with floater, dimensions (W x H x D): 65 x 405 x 335 mm |
| Dimensions (L x W x H) | 65 x 405 x 335 mm |
| For Use With (Application) | Collection of Flammable Liquid Waste |
| Includes | Canister with Floater |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?