Learn More
BPS Safety cabinet, Class II, BPS eco safe Plus Cytotoxic, 3ft
Brand:  BPS O000211A0100
Product Code. 15414812
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Plus Cytotoxic Class II Safety Cabinet conforms to DIN12980 and EN12469. External structure of epoxy powder coated steel, AISI-304L stainless steel back wall, safety glass side windows, motorised angled front sash, AISI-316L stainless steel one piece work surface. Three H14 HEPA filters, microprocessor based monitoring system with audio-visual alarm system. Double fan with automatic and independent airflow speed regulator. One vacuum tap, one fuel gas tap with solenoid valve, DEHS 100% port, one electrical socket for TCS 3 and TCS 4, two for TCS 6 included as standard.
  • External dimensions: W1045 x H2345 x D860mm
  • Motorised, sliding and hinged front window
  • Glass side windows
  • 304 stainless steel rear wall
  • 316 stainless steel single-piece worktop
  • Dual fan system with single exhaust H14 HEPA filters
  • Microprocessor control
  • Supplied with two electrical outlets, 1 x vacuum tap, 1 x gas tap
  • Half speed fan reduction for overnight set-back operation