missing translation for 'onlineSavingsMsg'
Learn More

PSF Progressive Safety™ Safety boots Quick Release 325mm Foundry Boot

Product Code. 12999393
Change view
Click to view available options
Size:
5
6
7
8
9
10
12
Unit Size:
1 pair
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12999393 7 1 pair
12979393 5 1 pair
12989393 6 1 pair
12909403 8 1 pair
12919403 9 1 pair
12949393 10 1 pair
12969393 12 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 12999393 Supplier PSF Progressive Safety™ Supplier No. 1177

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rubber nitrile sole

  • 200 joule toe protection
  • Manufactured to BS4676
  • Rubber nitrile sole
  • Sole resists oils acids, alkalis & heat to 300°C
  • Black
  • PSF Special Hazard

Specifications

Size 7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.