Learn More
WLD Tec™ Safety Adapter 360
Product Code. 11704179
Change view
Click to view available options
For Use With (Equipment):
Flame100
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
11704179 Flame100 Each
1 options
Product Code. 11704179 Supplier WLD Tec™ Supplier No. 2.000.701
Only null left
Add to basket
Add to basket
Safety adapter 360 with integrated pressure regulator for flame 100.
  • For Flame 100.
  • With integrated pressure regulator
  • For cartridges CV 360, 52 g from Camping Gaz.
  • Continuous cartridge operation 65 minutes.
Specifications
For Use With (Equipment) Flame100