Learn More
MEDICOM Safemask™ Fit+Pro FFP3 NR Valve Mask
Product Code. 31781311 Shop All Medicom Products
Change view
Click to view available options
Size:
One Size
Unit Size:
Pack of 144
Product Code. Size Unit Size
31781311 One Size Pack of 144
1 missing translation for 'options'
Product Code. 31781311 missing translation for 'mfr' MEDICOM missing translation for 'supplierNo' M63110V
  • ONLINE EXCLUSIVE PRICE
    £379.43
    £318.11 / Pack of 144
    Save £61.32
    16% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
SafeMask Fit+Pro FFP3 valved mask protects against toxic aerosols and high concentrations, providing enhanced breathing comfort.
  • Folded 3-panel design adapts to various face shapes and sizes
  • Sealing foams on both the upper and lower panels ensure an ideal seal while providing unmatched comfort for the wearer
  • Easy to put on and adjust thanks to sliding elastic straps and a tightening ring
  • The exhalation valve effectively expels heat and humidity, reducing the risk of fogging on the wearer's glasses
  • Its individual packaging helps control the risk of contamination
Specifications
Color White
Design Type Flat-fold
Disposable Single-use
Filter Type FFP3
Material Polypropylene
Product Type General Purpose Particulate Respirator
Size One Size
With Exhalation Valve Yes