missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ SafeLock™ Cover for Electrodes
Shop All Mettler Toledo Products
Click to view available options
For Use With (Equipment):
pH electrodes
Unit Size:
Each
Description
- White in color
- Pack of 5
Specifications
Specifications
| Color | White |
| Description | SafeLock white (5 pcs.) |
| For Use With (Application) | Closure of refill holes |
| For Use With (Equipment) | pH electrodes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction