Learn More
DWK Life Sciences Safeguarding, for safety standard ground sockets
Component of the system for the safe loosening of stuck ground joints.
Brand: DWK Life Sciences 286217302
Additional Details : Weight : 0.18000kg
Description
Polyamide tension rings have very good sliding properties. They help to safely loosen fixed joints through even distribution of force.
- By means of an even force distribution, stuck joints can be safely loosened.
- Due to their material properties, pull rings have very good sliding properties.
Specifications
10/Pk. | |
To secure the security grinding | |
Safeguard | |
NS 34/35 |