Learn More
DWK Life Sciences Safeguarding, for safety standard ground sockets
Component of the system for the safe loosening of stuck ground joints.
Brand:  DWK Life Sciences 286216709
Code : NEW
Additional Details : Weight : 0.00085kg
 View more versions of this product
Product Code. 10161741
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Polyamide tension rings have very good sliding properties. They help to safely loosen fixed joints through even distribution of force.
  • By means of an even force distribution, stuck joints can be safely loosened.
  • Due to their material properties, pull rings have very good sliding properties.
Specifications
Specifications
10/Pk.
To secure the security grinding
Safeguard
NS 45/40