Learn More
Eppendorf™ Safe-Lock Tubes, Forensic DNA Grade
Provides protection against unintentional opening during incubation and storage. Eppendorf™ Safe-Lock Tubes, Forensic DNA Grade can withstand forces up to 30,000 x g.
Brand: Eppendorf™ 0030123611
22.80 GBP valid until 2024-06-30
Use promo code "22546" to get your promotional price.
Additional Details : Weight : 0.65340kg
Description
- Hinged lid prevents unintentional opening during incubation and storage
- Polypropylene provides high resistance to chemicals, mechanical stress and temperature extremes
- Manufactured without mold release agents, softeners or biocides (materials which could have negative effect on bioassays)
- g-Safe™ Centrifugation stability up to 30,000 x g prevents sample loss due to tube breakage
- Provides extra safety when working with hazardous samples
- Precise lid sealing for minimal evaporation rates during long-term storage
- Large frosted lid, surface on the side for easy labeling
- Functions from -86° to 100°C
- Autoclavable (121°C, 20 min)
Specifications
30,000 x g | |
Colorless | |
Yes | |
Clear | |
Incubation and storage | |
Bag | |
Forensic DNA Grade |
Hinged lid | |
Polypropylene | |
1.5 mL | |
Polypropylene | |
10 x 50 | |
−86°C to 100°C | |
Yes |