missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Safe Imager™ 2.0 Blue-Light Transilluminator

Product Code. p-7180550
Change view
Click to view available options
Electrical Requirements:
Plug type: UK
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Electrical Requirements unitSize
13193102 Plug type: UK Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13193102 Supplier Invitrogen™ Supplier No. G6600UK

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Designed for viewing stained gels on the laboratory benchtop, or within a gel documentation system.

  • Supplied with a Safe Imager amber filter unit, a power cord, and Safe Imager viewing glasses.
  • Light from the LED source inside the transilluminator passes through a blue filter, producing a single-intensity signal at approximately 470 nm, effective for the excitation of SYBR Safe DNA gel stain, and many other nucleic acid and protein stains, such as SYBR Gold, SYBR Green I and II, SYPRO Ruby, SYPRO Orange, and Coomassie Fluor Orange stains
  • Sensitivity with this instrument is comparable to that of standard UV transilluminators
  • Does not produce UV light and does not require UV-protective equipment during use
  • Green Features: Energy efficient; Label or Dye: SYBR Safe
  • Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY

Specifications

Product Type Transilluminator
For Use With (Equipment) Safe Imager™
Dimensions (L x W x H) 29.5 x 32.5 x 6.5 cm
Electrical Requirements Plug type: UK
Content And Storage • Safe Imager
• Amber filter unit
• Power cord
• Safe Imager viewing glasses

Shipping Condition Room Temperature
Lighting Type LED Blue
Quantity 1 unit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.