Learn More
Hirschmann™ Safe-Cone Filters for Labopette™
Product Code. 10173913
Change view
Click to view available options
Volume (Metric):
Var: 10-100/20-200 μL, Fixed: 100/200 μL
Var: 10-500/50-1,000/500-5,000/50-1200 μL, 100-1000 μL, Fixed: 250/500/1000 μL
Var: 1000-5000 μL, Fixed: 2000/5000 μL
Var: 2-20/5-50 μL, Fixed 20/25/50 μL
Unit Size:
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10173913 Var: 10-500/50-1,000/500-5,000/50-1200 μL, 100-1000 μL, Fixed: 250/500/1000 μL Pack of 50
10233151 Var: 1000-5000 μL, Fixed: 2000/5000 μL Pack of 50
10012223 Var: 2-20/5-50 μL, Fixed 20/25/50 μL Pack of 50
10052173 Var: 10-100/20-200 μL, Fixed: 100/200 μL Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10173913 Supplier Hirschmann™ Supplier No. 9470011
Only null left
Add to basket
Add to basket
For the prevention of cross contamination of liquid samples or vapours when pipetting radioactive, biohazardous or corrosive samples.
  • Provides the decisive advantage when protecting pipettes and samples against contamination
  • Extends the service life of pipettes
  • Provides excellent protection for the internal components if replaced regularly
  • Can serve as additional protection against penetration of liquids into the interior of the pipette
  • For use with a range of Labopette™ models
Specifications
Type Safety Filter
Volume (Metric) Var: 10-500/50-1,000/500-5,000/50-1200 μL, 100-1000 μL, Fixed: 250/500/1000 μL
For Use With (Application) Protection against cross-contamination
For Use With (Equipment) Labopette models 9480104, 9480105, 9480106, 9475409, 9475508, 9475509, 9475510, 9480800