missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAFB2 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11642-1-AP-20UL
This item is not returnable.
View return policy
Description
SAF-B2 (scaffold attachment factor B2) is a nuclear matrix-associated protein that binds to matrix- or scaffold-associating regions (MARs or SARs) on DNA and interacts with RNA polymerase II and serine-/arginine-rich RNA processing factors (SR proteins). SAF-B2, and the related SAF-B, bind to the estrogen receptor (ER) and are expressed in several breast cancer cell lines at varying levels. Subsequently, SAF-B and SAF-B2 may play a role in breast cancer by mediating cellular proliferation and division. Unlike SAF-B, which is exclusively nuclear, SAF-B2 is found in the cytoplasm as well as the nucleus.Specifications
| SAFB2 | |
| Polyclonal | |
| Unconjugated | |
| SAFB2 | |
| KIAA0138, SAF B2, SAFB2, scaffold attachment factor B2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 301126, 9667 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q14151 | |
| SAFB2 | |
| SAFB2 Fusion Protein Ag2230 | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction