missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SAFB Antibody (6F7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27940
This item is not returnable.
View return policy
Description
SAFB Monoclonal antibody specifically detects SAFB in Human,Monkey,Orangutan,Primate samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| SAFB | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 - 1:3000, Immunohistochemistry 10 μg/mL, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation | |
| Q15424 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Monkey, Orangutan, Primate | |
| Purified |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 6F7 | |
| Unconjugated | |
| PBS | |
| glutathione S-transferase fusion protein, HAP, heat-shock protein (HSP27) estrogen response element and TATA box-bindingprotein, HETDKFZp779C1727, Hsp27 ERE-TATA binding protein, HSP27 ERE-TATA-binding protein, HSP27 estrogen response element-TATA box-binding protein, SAF-B, SAF-B1, SAFB1SAB-B1, scaffold attachment factor B, scaffold attachment factor B1 | |
| SAFB monoclonal antibody was raised against amino acids 345 - 357 of SAFB (Human). | |
| 0.05 mg | |
| DNA Repair | |
| 6294 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction