missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SAE1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583666
This item is not returnable.
View return policy
Description
Immunogen sequence: KGLTMLDHEQ VTPEDPGAQF LIRTGSVGRN RAEASLERAQ NLNPMVDVKV DTEDIEKKPE SFFTQFDAVC LTCCSRD Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 95%.
Posttranslational modification of proteins by the addition of the small protein SUMO (see SUMO1), or sumoylation, regulates protein structure and intracellular localization. SAE1 and UBA2. form a heterodimer that functions as a SUMO-activating enzyme for the sumoylation of proteins (Okuma et al., 1999).
Specifications
| SAE1 | |
| Polyclonal | |
| Unconjugated | |
| SAE1 | |
| 2400010M20Rik; 2610044L12Rik; activator of SUMO1; AL033372; AOS1; AW743391; D7Ertd177e; FLJ3091; HSPC140; Sae1; sentrin/SUMO-activating protein AOS1; Sua1; SUMO-1 activating enzyme E1 N subunit; SUMO1 activating enzyme subunit 1; SUMO-1 activating enzyme subunit 1; SUMO-activating enzyme subunit 1; SUMO-activating enzyme subunit 1, N-terminally processed; ubiquitin-like (sentrin) activating enzyme E1A; ubiquitin-like 1 (sentrin) activating enzyme E1A; ubiquitin-like 1 (sentrin) activating enzyme subunit 1; ubiquitin-like 1-activating enzyme E1A; ubiquitin-like protein SUMO-1 activating enzyme; Ubl1a1; Uble1a | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10055, 308384, 56459 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6AXQ0, Q9R1T2, Q9UBE0 | |
| SAE1 | |
| Recombinant protein corresponding to Human SAE1. Recombinant protein control fragment (Product #RP-99109). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction