missing translation for 'onlineSavingsMsg'
Learn More

Sacubitril, MedChemExpress

Product Code. 16486488
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16486488

Brand: Medchem Express HY15407

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sacubitril (AHU-377) is a potent and orally active NEP (neprilysin) inhibitor with an IC50 of 5 nM. Sacubitril is a component of the heart failure medicine LCZ696. Sacubitril can be used for the research of heart failure, hypertension and COVID-19.

Sacubitril (AHU-377) is a potent and orally active NEP (neprilysin) inhibitor with an IC50 of 5 nM. Sacubitril is a component of the heart failure medicine LCZ696. Sacubitril can be used for the research of heart failure, hypertension and COVID-19.

Specifications

Chemical Name or Material Sacubitril
Physical Form Oil
CAS 149709-62-6
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C24H29NO5
For Use With (Application) COVID-19-immunoregulation
Synonym AHU-377
Solubility Information DMSO : 100 mg/mL (243.02 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (1.63 mM; Need ultrasonic)
SMILES O=C(CCC(O)=O)N[C@H](CC1=CC=C(C2=CC=CC=C2)C=C1)C[C@@H](C)C(OCC)=O
Molecular Weight (g/mol) 411.49
Formula Weight 411.49
Percent Purity 99.18%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.