missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Sacsin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579951
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat brain tissue, mouse brain tissue, human Hela cell. IHC: mouse brain tissue, rat brain tissue.
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Kruppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. ANKZF1 (ankyrin repeat and zinc finger domain containing 1), also known as ZNF744, is 726 amino acids in length and lacks a KRAB domain but contains one C2H2-type zinc finger and two ANK repeats. The gene encoding ANKZF1 localizes to chromosome 2.
Specifications
| Sacsin | |
| Polyclonal | |
| Unconjugated | |
| SACS | |
| A230052M14; ARSACS; DKFZp686B15167; dnaJ homolog subfamily C member 29; DNAJC29; E130115J16Rik; Kiaa0730; mKIAA0730; PPP1R138; protein phosphatase 1, regulatory subunit 138; RGD1305416; Sacs; Sacsin; sacsin molecular chaperone; spastic ataxia of Charlevoix-Saguenay (sacsin); SPAX6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26278, 305940, 50720 | |
| -20°C | |
| Lyophilized |
| ELISA, Immunohistochemistry (Paraffin) | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| Q9JLC8, Q9NZJ4 | |
| SACS | |
| E. coli-derived human Sacsin recombinant protein (Position: E3709-L3909). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction