missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SACM1L Polyclonal Antibody

SACM1L Polyclonal Antibody
Brand: Invitrogen PA567080
This item is not returnable.
View return policy
Description
SACM1L Polyclonal Antibody for ICC/IF
Specifications
| SACM1L | |
| Polyclonal | |
| Unconjugated | |
| Sacm1l | |
| Kiaa0851; mKIAA0851; phosphatidylinositide phosphatase SAC1; phosphatidylinositol-3-phosphatase SAC1; phosphatidylinositide phosphatase SAC1; phosphatidylinositol-4-phosphate phosphatase; polyphosphatidylinositol phosphatase; SAC1; SAC1 (suppressor of actin mutations 1, homolog)-like (S. cerevisiae); SAC1 like phosphatidylinositide phosphatase; SAC1 suppressor of actin mutations 1 like; SAC1 suppressor of actin mutations 1-like (yeast); Sac1p; SACM1L; suppressor of actin 1; suppressor of actin mutations 1-like protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22908 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NTJ5 | |
| Sacm1l | |
| Recombinant Human SACM1L. Recombinant protein control fragment (Product #RP-105492). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction