Description
- Quick and easy to apply and remove
- Ratchet mechanism and secondary positive lock for double security
- Robust
- Reusable
- Made from Polyamide (PA)
- Simply close sack between the jaws of the Sack Clip
Specifications
Specifications
| Material | Polyamide |
| Size | 100 mm jaw length |
| Color | White |
| Length (Metric) | 250 mm |