missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Saccharomyces cerevisiae ZIP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117585
This item is not returnable.
View return policy
Description
This antibody has been tested in direct-ELISA.
ZIP1, also known as ZIRTL (zinc-iron regulated transporter-like), is the first mammalian member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. ZIP1 expression is markedly downregulated in a number of cancerous tissues and is thought to function as a tumor suppressor gene in prostate cancer. More recent studies have shown that overexpression of ZIP1 and concomitant increased levels of intracellular zinc in PC-3 cells cause a significant inhibition of NF-kappa-B, leading to down-regulation of the antiapoptotic protein Bcl-2, Bcl-XL, and the apoptosis inhibitor XIAP, reducing the malignant potential of prostate cancer cells in vitro and in vivo.
Specifications
| Saccharomyces cerevisiae ZIP1 | |
| Polyclonal | |
| Unconjugated | |
| ZIP1 | |
| CGI-08; CGI-71; IRT1; mZIP1; SLC39A1; solute carrier family 39 (zinc transporter), member 1; solute carrier family 39 (zinc transporter), member 3; solute carrier family 39 member 1; Zinc transporter ZIP1; Zinc-iron-regulated transporter-like; Zip1; ZIP-1; ZIRTL; Zrt- and Irt-like protein 1; Zrt/Irt-like protein 1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 851879 | |
| -20°C or -80°C if preferred | |
| Liquid |
| Western Blot | |
| 2.95 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P31111 | |
| ZIP1 | |
| Recombinant Saccharomyces cerevisiae Synaptonemal complex protein ZIP1 protein (1-227aa). | |
| 100 μL | |
| Primary | |
| Yeast | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction