missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ SABM™ Small Airway Epithelial Cell Growth Basal Medium

Product Code. 11665420
Click to view available options
Quantity:
500 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11665420

Brand: Lonza™ CC3119

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The basal medium component in the full SAGM™ Small Airway Epithelial Cell Growth Medium BulletKit™ (catalog number CC-3118), which consists of basal and supplements packaged together.

  • Basal medium.
  • Component in the full SAGM™ Small Airway Epithelial Cell Growth Medium BulletKit™ (catalog number CC-3118) which consists of basal and supplements packaged together.
  • Serum-free and optimized to grow human small airway epithelial cells (SAEC).
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Airway Cells
Product Type Basal Medium
Quantity 500 mL
Format 500 mL Bottle
pH 7.4 to 7.6
Color Red
Content And Storage Store in dark at 2 to 8°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.