missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SAB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530315
This item is not returnable.
View return policy
Description
Recommended positive controls: IMR32. Predicted reactivity: Mouse (87%), Rat (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Inhibits the auto- and transphosphorylation activity of BTK. Plays a negative regulatory role in BTK-related cytoplasmic signaling in B-cells. May be involved in BCR-induced apoptotic cell death.
Specifications
| SAB | |
| Polyclonal | |
| Unconjugated | |
| SH3BP5 | |
| AI606498; Sab; SH3 binding protein; SH3 domain binding protein 5; SH3 domain-binding protein 5; SH3 domain-binding protein that preferentially associates with BTK; SH3BP5; SH3BP-5; SH3-domain binding protein 5 (BTK-associated); Vascular endothelial cell-specific protein 18; Vesp18 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9467 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| O60239 | |
| SH3BP5 | |
| Recombinant protein encompassing a sequence within the center region of human SH3BP5. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction