missing translation for 'onlineSavingsMsg'
Learn More

Sa Cas9 Rabbit anti-Bacteria, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 26927-1-AP-20UL

 View more versions of this product

Product Code. 16885255

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody detects the Cas9 of Staphylococcus aureus (SaCas9). It does not detect the SpCas9.

Cas9 (CRISPR associated protein 9) is an RNA-guided DNA endonuclease enzyme associated with the CRISPR (Clustered Regularly Interspaced Short Palindromic Repeats) adaptive immunity system in Streptococcus pyogenes (Sp).
Specifications

Specifications

Sa Cas9
Polyclonal
Unconjugated
Cas9, Sa Cas9, SaCas9
Sa Cas9 Fusion Protein Ag25669
20 μL
Primary
-20°C
Liquid
Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Rabbit
Antigen Affinity Chromatography
RUO
Bacteria
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.