missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sa Cas9 Rabbit anti-Bacteria, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26927-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody detects the Cas9 of Staphylococcus aureus (SaCas9). It does not detect the SpCas9.
Cas9 (CRISPR associated protein 9) is an RNA-guided DNA endonuclease enzyme associated with the CRISPR (Clustered Regularly Interspaced Short Palindromic Repeats) adaptive immunity system in Streptococcus pyogenes (Sp).Specifications
| Sa Cas9 | |
| Polyclonal | |
| Unconjugated | |
| Cas9, Sa Cas9, SaCas9 | |
| Sa Cas9 Fusion Protein Ag25669 | |
| 150 μL | |
| Primary | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction