missing translation for 'onlineSavingsMsg'
Learn More

SA 47, MedChemExpress

Product Code. 16469937
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16469937 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16469937 Supplier Medchem Express Supplier No. HY18080
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress SA 47 is a selective and potent inhibitor of fatty acid amide hydrolase (FAAH) and carbamate.

SA 47 is a selective and potent inhibitor of fatty acid amide hydrolase (FAAH) and carbamate.

Specifications

Chemical Name or Material SA 47
CAS 792236-07-8
Color White
Purity Grade Notes Research
Molecular Formula C17H26N4O3
Quantity 5 mg
SMILES CNC(COC(NCCC(CC1)CCN1C2=CC=CC(C)=N2)=O)=O
Molecular Weight (g/mol) 334.41
Formula Weight 334.41
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.