missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SA 4503 dihydrochloride
GREENER_CHOICE

Product Code. 30002026
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30002026

Brand: Tocris Bioscience™ 4951/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective σ1 receptor agonist

Selective σ1 receptor agonist (IC50 = 17.4 nM). Exhibits selectivity for σ1 over σ2 receptors. Inhibits angiotensin II-induced cardiomyocyte hypertrophy in vitro and attenuates myocardial hypertrophy in vivo. Enhances brain plasticity and sensorimotor function in a rat model of experimental stroke.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 165377-44-6
Molecular Formula C23H34Cl2N2O2

Specifications

Quantity 10 mg
Formula Weight 441.43
Physical Form Solid
Color White
Purity >98%
Molecular Weight (g/mol) 441.437
Chemical Name or Material SA 4503 dihydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.