Learn More
Socorex™ Acura™ Electro 926 Micropipette
Product Code. 11829213
Change view
Click to view available options
Volume (Metric):
0.1 to 2.0 μL
0.5 to 10 μL
0.5 to 10 μL (Ultra)
1.0 to 20 μL
10 to 200 μL
2.5 to 50 μL
50 to 1000 μL
Type:
Electronic Micropipette
Electronic Micropipette Kit
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Type Unit Size
11829213 0.5 to 10 μL (Ultra) Electronic Micropipette Each
11829223 10 to 200 μL Electronic Micropipette Each
11849223 50 to 1000 μL Electronic Micropipette Each
11839223 10 to 200 μL Electronic Micropipette Kit Each
11809213 0.1 to 2.0 μL Electronic Micropipette Each
11849213 0.5 to 10 μL Electronic Micropipette Each
11869213 1.0 to 20 μL Electronic Micropipette Each
11889213 2.5 to 50 μL Electronic Micropipette Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11829213 Supplier Socorex™ Supplier No. 926.0010
 Restricted Item
In Stock
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Lightweight and ergonomically friendly, this electronic single-channel micropipette comes with a QC certificate, Qualitips™ pipette tip samples and operating instructions. Socorex™ Acura™ Electro 926 Micropipette charges quickly and has a long working autonomy.
  • Exceptional, user-friendly ergonomics.
  • Lightweight.
  • Easy, intuitive, self-teaching program.
  • Large display, reversible right/left reading.
  • Microprocessor-controlled motor.
  • Swift-interchange, rapidly charging, long-working, rechargeable battery pack.
  • Reduced shaft length and conical end improve drivability and access to microtubes and microplates.
  • Includes pipetting cycle counter.
  • Optional volumetric modules.
Spezifikation
Accuracy <±1.2% - <±0.6%
Battery Type 4.8V NiMH
Certifications/Compliance EN ISO 8655, GMP, GLP, NCCLS. IVD 98/79 EEC (in vitro diagnostic medical devices), ED 89/336 EEC, 91/263 EEC, 92/31 EEC, 93/68 EEC, FCCCFR47, part 15 (electromagnetic compatibility), ED 73/23 EEC, 93/68EEC, IEC 61010-1 (electrical safety), ED 2011/65/EC Restriction of Hazardous Substances (RoHS)
Model 926
Number of Channels 1
Product Line Acura Electro
Programmability Self-teaching
Type Electronic Micropipette
Volume (Metric) 0.5 to 10 μL (Ultra)
Display Right/Left LCD
Includes QC certificate, Qualitips™ pipette tip samples and operating instructions.
Increments 0.05μL
Precision <1.5% - <0.35%
Quantity 1 Ea.
Recharge Time <1.5 Hours
Warranty 24 Months
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.