missing translation for 'onlineSavingsMsg'
Learn More

S63845, MedChemExpress

Product Code. 16415887
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16415887

Brand: Medchem Express HY100741

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress S63845 is a potent and selective myeloid cell leukemia 1 (MCL1) inhibitor with a Kd of 0.19 nM for human MCL1.

S63845 is a potent and selective myeloid cell leukemia 1 (MCL1) inhibitor with a Kd of 0.19 nM for human MCL1.

Specifications

Chemical Name or Material S63845
Physical Form Solid
CAS 1799633-27-4
Color Light Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C39H37ClF4N6O6S
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 33.33 mg/mL (40.19 mM; Need ultrasonic)
SMILES CC(C(Cl)=C(OCCN1CCN(C)CC1)C=C2)=[C@@]2[C@]3=C(C4=CC=C(F)O4)SC5=NC=NC(O[C@H](CC6=C(OCC7=CC=NN7CC(F)(F)F)C=CC=C6)C(O)=O)=C53
Molecular Weight (g/mol) 829.26
Formula Weight 829.26
Percent Purity 96.47%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.