missing translation for 'onlineSavingsMsg'
Learn More

S5a/Angiocidin Mouse anti-Human, Clone: CPTC-PSMD4-3, Novus Biologicals™

Product Code. 18383913 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18383913 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18383913 Supplier Novus Biologicals Supplier No. NBP30725020UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

S5a/Angiocidin Monoclonal specifically detects S5a/Angiocidin in Human samples. It is validated for Western Blot, Protein Array.
TRUSTED_SUSTAINABILITY

Specifications

Antigen S5a/Angiocidin
Applications Western Blot, Peptide Array
Classification Monoclonal
Clone CPTC-PSMD4-3
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Host Species Mouse
Immunogen Recombinant full-length human S5a/Angiocidin protein
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Neuroscience, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5710
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.