missing translation for 'onlineSavingsMsg'
Learn More

S5a/Angiocidin Antibody [PerCP], Novus Biologicals™

Product Code. 30732330 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732330 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732330 Supplier Novus Biologicals Supplier No. AF8036C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

S5a/Angiocidin Polyclonal antibody specifically detects S5a/Angiocidin in Human,Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen S5a/Angiocidin
Applications Western Blot
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Gene Alias 26S proteasome non-ATPase regulatory subunit 4;AF;AF-1;ASF;MCB1;PSMD4;pUB-R5;Rpn10;S5A
Host Species Sheep
Immunogen E. coli-derived recombinant human S5a/Angiocidin, Met1-Lys377, Accession # P55036
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5710
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.