missing translation for 'onlineSavingsMsg'
Learn More

S55746, MedChemExpress

Product Code. 16447627
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16447627

Brand: Medchem Express HY117288

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress S55746 (BCL201) is a potent, orally active and selective BCL-2 inhibitor, with a Ki of 1.3 nM and a Kd of 3.9 nM. S55746 (BCL201) has antitumor activity with low toxicity.

S55746 (BCL201) is a potent, orally active and selective BCL-2 inhibitor, with a Ki of 1.3 nM and a Kd of 3.9 nM. S55746 (BCL201) has antitumor activity with low toxicity.

Specifications

Chemical Name or Material S55746
CAS 1448584-12-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C43H42N4O6
Quantity 5 mg
Synonym BCL201
Solubility Information DMSO : 50 mg/mL (70.34 mM; Need ultrasonic)
SMILES O=C(C1=C2CCCCN2C(C3=C(C(N4CC5=C(C=CC=C5)C[C@H]4CN6CCOCC6)=O)C=C(OCO7)C7=C3)=C1)N(C8=CC=C(O)C=C8)C9=CC=CC=C9
Molecular Weight (g/mol) 710.82
Formula Weight 710.82
Percent Purity 98.93%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.