missing translation for 'onlineSavingsMsg'
Learn More

S55746, MedChemExpress

Product Code. 16481127
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481127 50 mg 50mg
16465656 100 mg 100mg
16447627 5 mg 5mg
16434878 10 mg 10mg
16413276 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16481127 Supplier Medchem Express Supplier No. HY117288
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress S55746 (BCL201) is a potent, orally active and selective BCL-2 inhibitor, with a Ki of 1.3 nM and a Kd of 3.9 nM. S55746 (BCL201) has antitumor activity with low toxicity.

S55746 (BCL201) is a potent, orally active and selective BCL-2 inhibitor, with a Ki of 1.3 nM and a Kd of 3.9 nM. S55746 (BCL201) has antitumor activity with low toxicity.

Specifications

Chemical Name or Material S55746
CAS 1448584-12-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C43H42N4O6
Quantity 50 mg
Synonym BCL201
Solubility Information DMSO : 50 mg/mL (70.34 mM; Need ultrasonic)
SMILES O=C(C1=C2CCCCN2C(C3=C(C(N4CC5=C(C=CC=C5)C[C@H]4CN6CCOCC6)=O)C=C(OCO7)C7=C3)=C1)N(C8=CC=C(O)C=C8)C9=CC=CC=C9
Molecular Weight (g/mol) 710.82
Formula Weight 710.82
Percent Purity 98.93%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.