missing translation for 'onlineSavingsMsg'
Learn More

S55746 hydrochloride, MedChemExpress

Product Code. 16491127
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491127 50 mg 50mg
16475656 100 mg 100mg
16457627 5 mg 5mg
16444878 10 mg 10mg
16423276 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16491127 Supplier Medchem Express Supplier No. HY117288A
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress S55746 hydrochloride (BCL201 hydrochloride) is a potent, orally active and selective BCL-2 inhibitor, with a Ki of 1.3 nM and a Kd of 3.9 nM. S55746 hydrochloride (BCL201 hydrochloride) has antitumor activity with low toxicity.

S55746 hydrochloride (BCL201 hydrochloride) is a potent, orally active and selective BCL-2 inhibitor, with a Ki of 1.3 nM and a Kd of 3.9 nM. S55746 hydrochloride (BCL201 hydrochloride) has antitumor activity with low toxicity.

Specifications

Chemical Name or Material S55746 hydrochloride
CAS 1448525-91-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C43H43ClN4O6
Quantity 50 mg
Synonym BCL201 hydrochloride
Solubility Information DMSO : 200 mg/mL (267.64 mM; Need ultrasonic)
SMILES O=C(C1=C2CCCCN2C(C3=C(C(N4CC5=C(C=CC=C5)C[C@H]4CN6CCOCC6)=O)C=C(OCO7)C7=C3)=C1)N(C8=CC=C(O)C=C8)C9=CC=CC=C9.Cl
Molecular Weight (g/mol) 747.28
Formula Weight 747.28
Percent Purity 98.69%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.