missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ S3700 Multicolor and Cut, Sign and Label Printer
Color your way to an improved workplace with the S3700 Sign and Label Printer. With multicolor print and shape cutting capabilities, this printer creates impactful identification that fits your application and workspace needs.
POA
Specifications
Product Type | Printer |
---|---|
Color | Blue, Gray |
Display Type | Color LCD Touchscreen |
For Use With (Equipment) | Brady Labels |
Material | Plastic |
Description
- Color your way to an improved workplace with the S3700 Sign and Label Printer.
- With multicolor print and shape cutting capabilities, this printer creates impactful identification that fits your application and workspace needs.
- Multicolor print capabilities make it ideal for creating impactful safety signs, lockout tags, warning labels and more in a single pass
- Built in XY-cutter allows for the creation of die-cut shapes and text that gives the flexibility to choose where to place new messaging
- The built-in keyboard and touchscreen interface make label creation a breeze
- Label creation is even faster with the quick setup and print speeds that come with print smart technology from Brady
- Prints on a variety of material options so you can create signs and labels for a variety of applications
- Ships with Brady Workstation Basic Label Design software suite, which will enable you to immediately design professional labels.
Specifications
Printer | |
Color LCD Touchscreen | |
Plastic | |
14.25 in. | |
361.95 mm | |
11 in. | |
Multicolor, Monocolor | |
S3700 Safety and Facility Printer, Brady Workstation Basic Suite, Quick Start Guide, Power Cord, USB Cord, Mouse, Cleaning Kit (PCK-6), USB Flash Drive | |
S3700 | |
300 dpi | |
cULus, CE, UL-AR and S Mark, UL NOM CoC, RCM Mark, EAC, KC, CCC, RoHS, BIS, BSMI, LOA, SABS | |
1 Ea. | |
AC Adaptor |
Blue, Gray | |
Brady Labels | |
Thermal Transfer | |
19 in. | |
482.60 mm | |
279.40 mm | |
155.19 x 86.87 mm | |
512 MB | |
127.00 mm/sec. | |
16.74 kg | |
Asset tracking, Cold temperature labelling, Component and equipment labelling, Electrical labelling, Facility identification, General identification, Hazard communication identification | |
1 year | |
110/240 V |