missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S2I-201, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | 2-Hydroxy-4-[(([(4-methylphenyl)sulfonyl]oxy)acetyl)amino]benzoic Acid |
| CAS | 501919-59-1 |
| Molecular Formula | C16H15NO7S |
| Quantity | 10 mg |
| Synonym | NSC 74859, S2I-201 |
| InChI Formula | InChI=1S/C16H15NO7S/c1-10-2-5-12(6-3-10)25(22,23)24-9-15(19)17-11-4-7-13(16(20)21)14(18)8-11/h2-8,18H,9H2,1H3,(H,17,19)(H,20,21) |
| SMILES | Cc1ccc(cc1)S(=O)(=O)OCC(=O)Nc2ccc(C(=O)O)c(O)c2 |
| IUPAC Name | 2-hydroxy-4-[[2-(4-methylphenyl)sulfonyloxyacetyl]amino]benzoic acid |
| Molecular Weight (g/mol) | 365.36 |
| Formula Weight | 365.0569 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction