missing translation for 'onlineSavingsMsg'
Learn More

Elma™ Lab Clean S20 Concentrate

Product Code. 11882681
Click to view available options
Volume (Metric):
2.5 L
10 L
Unit Size:
10 L
2.5 L
This item is not returnable. View return policy

Product Code. 11882681

Brand: Elma™ 8000191

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Suitable for the acidic fundamental cleaning of dental and medical instruments, labware and workpieces made from stainless steel, glass, ceramic and plastic. Elma™ Lab Clean S20 Concentrate is liquid-, acid- and foam-inhibited for dip and splash cleaning.

  • Removes mineral contaminations like lime scales and other Ca- and Mg-bound contaminations, metal oxides, mineral oils and greases, fingerprints and dust
  • Brightens metallic surfaces tarnished (but not temper colours)
  • Not suitable for aluminum, magnesium and alloys of light metals

Specifications

Product Type Lab Clean S20 Cleaner
Form Liquid
pH <1
Certifications/Compliance Annex VII A, EC 648/2004, UN 1805
Composition Non-ionic surfactants 15 to 30%, Phosphoric acid >30%
Concentration Ultrasonic bath ∽1 to 2 vol% at 50 to 80°C and splash cleaning ∽0.5 vol%, well above 55°C only
Container Type Bottle
Content And Storage -5°C to 30°C
For Use With (Application) For dip and splash cleaning of instruments of labs and workshops applications
Volume (Metric) 2.5 L
compliance-icons compliance-icons
Product Identifier
  • elma lab clean S20 (ELC S20)
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308-IF exposed or concerned:.
Supplemental information
  • MIXTURE LIST-Contains : phosphoric acid, ethoxylated isotridecanol, (2-methoxymethylethoxy)-propanol, isotridecanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.