missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S1RA hydrochloride, MedChemExpress
Description
S1RA hydrochloride (E-52862 hydrochloride) is a potent and selective sigma-1 receptor(σ1R, Ki=17 nM) antagonist, showed good selectivity against σ2R (Ki > 1000 nM).
Specifications
Specifications
| Chemical Name or Material | S1RA hydrochloride |
| Physical Form | Solid |
| CAS | 1265917-14-3 |
| Color | Off-White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C20H24ClN3O2 |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Synonym | E-52862 hydrochloride |
| Solubility Information | DMSO : 100 mg/mL (267.47 mM; Need ultrasonic) ∣H2O : 16.67 mg/mL (44.59 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?