missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
S1P5/EDG-8 Monoclonal antibody specifically detects S1P5/EDG-8 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
Specifications
| Antigen | S1P5/EDG-8 |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Clone | 282503 |
| Conjugate | Janelia Fluor 585 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Edg-8, EDG8S1P receptor Edg-8, Endothelial differentiation G-protein-coupled receptor 8, endothelial differentiation, sphingolipid G-protein-coupled receptor, 8, S1P receptor 5, S1P5, sphingosine 1-phosphate receptor 5, sphingosine 1-phosphate receptor EDG8, Sphingosine 1-phosphate receptor Edg-8, sphingosine-1-phosphate receptor 5, SPPR-1, SPPR-2 |
| Host Species | Mouse |
| Immunogen | BaF3 mouse pro-B cell line transfected with human S1P5/EDG-8, Met1-Asp398, Accession # Q9H228 |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?