missing translation for 'onlineSavingsMsg'
Learn More

S1P5/EDG-8 Antibody (1196A) [Janelia Fluor™ 669], Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals FAB9084JF669

Product Code. 30171311

  • £476.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

S1P5/EDG-8 Monoclonal antibody specifically detects S1P5/EDG-8 in Mouse samples. It is validated for Flow Cytometry,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

S1P5/EDG-8
Monoclonal
Janelia Fluor 669
Edg-8, EDG8S1P receptor Edg-8, Endothelial differentiation G-protein-coupled receptor 8, endothelial differentiation, sphingolipid G-protein-coupled receptor, 8, S1P receptor 5, S1P5, sphingosine 1-phosphate receptor 5, sphingosine 1-phosphate receptor EDG8, Sphingosine 1-phosphate receptor Edg-8, sphingosine-1-phosphate receptor 5, SPPR-1, SPPR-2
Mouse S1P5/EDG-8 peptide corresponding to the N-terminal extracellular sequence, Accession # Q91X56
RUO
Primary
Mouse
Liquid
Flow Cytometry, Western Blot
1196A
50mM Sodium Borate
Rabbit
0.1 mL
GPCR
53637
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.